Product Details
- SNP ID
-
hCV9622232
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:163146563 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAATTTGAAAATGGAAACATTTGA[C/T]CCACAGTCTAGCAGCATAAATACAT
- Phenotype
-
MIM: 603276
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RGS5
PubMed Links
Gene Details
- Gene
- RGS5
- Gene Name
- regulator of G-protein signaling 5
There are no transcripts associated with this gene.
View Full Product Details