Product Details
- SNP ID
-
rs1803287
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:151983165 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGCGAACTGCTCATTTCTGCCT[A/G]CTTCTTTCCCTTCTGCTTCATGTGT
- Phenotype
-
MIM: 114085
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
S100A10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143276981] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- S100A10
- Gene Name
- S100 calcium binding protein A10
There are no transcripts associated with this gene.
View Full Product Details