Product Details

SNP ID
rs944682
Assay Type
Functionally tested
NCBI dbSNP Submissions
68
Location
Chr.1:152720112 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GGCCCCAGAACTGCAACACAGGATC[A/G]TCTGGCTGCTGTGAGAATTCGGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf68 PubMed Links

Gene Details

Gene
C1orf68
Gene Name
chromosome 1 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024679.2 591 Silent Mutation TCA,TCG S197S NP_001019850.1

View Full Product Details