Product Details

SNP ID
rs1058195
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:155195730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGAACTCCTTTTGGGAGCCAGA[A/G]AAGGGTCTGTGGTTGGCTGAGGGGC
Phenotype
MIM: 158340 MIM: 188062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR92B PubMed Links

Gene Details

Gene
MIR92B
Gene Name
microRNA 92b
There are no transcripts associated with this gene.

Gene
MUC1
Gene Name
mucin 1, cell surface associated
There are no transcripts associated with this gene.

Gene
THBS3
Gene Name
thrombospondin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252607.1 3038 UTR 3 NP_001239536.1
NM_001252608.1 3038 UTR 3 NP_001239537.1
NM_007112.4 3038 UTR 3 NP_009043.1
XM_006711498.3 3038 UTR 3 XP_006711561.1
XM_011509926.2 3038 UTR 3 XP_011508228.1
XM_011509927.2 3038 UTR 3 XP_011508229.1
XM_011509928.2 3038 UTR 3 XP_011508230.1
XM_011509929.1 3038 UTR 3 XP_011508231.1
XM_011509930.2 3038 UTR 3 XP_011508232.1
XM_011509931.1 3038 UTR 3 XP_011508233.1
XM_011509932.2 3038 UTR 3 XP_011508234.1
XM_011509933.1 3038 UTR 3 XP_011508235.1
XM_011509934.1 3038 UTR 3 XP_011508236.1
XM_011509935.1 3038 UTR 3 XP_011508237.1
XM_011509936.1 3038 UTR 3 XP_011508238.1
XM_017002206.1 3038 UTR 3 XP_016857695.1

View Full Product Details