Product Details

SNP ID
rs708742
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:226549806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCCTTCTGTTCCTGAGAAGTAG[A/G]GAGTGCGCCGATGTAATTCTGTGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STUM PubMed Links

Gene Details

Gene
STUM
Gene Name
stum, mechanosensory transduction mediator homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003665.3 Intron NP_001003665.1
XM_011544183.2 Intron XP_011542485.1

View Full Product Details