Product Details

SNP ID
rs755554
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:47410483 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTAGGAGACCCCAGGTCACAGAGT[C/G]TCTGAGATGGAGGGAGAGGATAGAA
Phenotype
MIM: 186852 MIM: 608735
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR4487 PubMed Links

Gene Details

Gene
MIR4487
Gene Name
microRNA 4487
There are no transcripts associated with this gene.

Gene
PSMC3
Gene Name
proteasome 26S subunit, ATPase 3
There are no transcripts associated with this gene.

Gene
SLC39A13
Gene Name
solute carrier family 39 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128225.2 Intron NP_001121697.1
NM_152264.4 Intron NP_689477.2
XM_006718381.3 Intron XP_006718444.1
XM_006718383.3 Intron XP_006718446.1
XM_006718384.2 Intron XP_006718447.1
XM_006718385.2 Intron XP_006718448.1
XM_011520466.1 Intron XP_011518768.1
XM_011520467.1 Intron XP_011518769.1
XM_011520468.2 Intron XP_011518770.1
XM_011520469.1 Intron XP_011518771.1
XM_011520470.1 Intron XP_011518772.1
XM_017018540.1 Intron XP_016874029.1
XM_017018541.1 Intron XP_016874030.1
XM_017018542.1 Intron XP_016874031.1

View Full Product Details