Product Details

SNP ID
rs1048908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98613578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTACCTTAGTCTAATCTCAATAA[C/T]GTCAAACTAATTCATATCTGAAATG
Phenotype
MIM: 609861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKBIP PubMed Links

Gene Details

Gene
IKBIP
Gene Name
IKBKB interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153687.3 1292 Intron NP_710154.1
NM_201612.2 1292 UTR 3 NP_963906.1
NM_201613.2 1292 UTR 3 NP_963907.1

View Full Product Details