Product Details

SNP ID
rs1465457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72012369 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGAAAACCAGAACCCCAAGTGAG[C/G]AGTGGGGATTTGGAATTCAGTCTGA
Phenotype
MIM: 126064
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DHODH PubMed Links

Gene Details

Gene
DHODH
Gene Name
dihydroorotate dehydrogenase (quinone)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001361.4 Intron NP_001352.2
XM_005255827.3 Intron XP_005255884.1
XM_005255829.3 Intron XP_005255886.1
XM_017022990.1 Intron XP_016878479.1

View Full Product Details