Product Details

SNP ID
rs1046156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:61853015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAAATGTTGTTATGTTAAACAT[C/T]ATGAGCAAATCTAAAGGTTCAACCA
Phenotype
MIM: 608879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPS13C PubMed Links

Gene Details

Gene
VPS13C
Gene Name
vacuolar protein sorting 13 homolog C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018088.2 12795 Intron NP_001018098.1
NM_017684.4 12795 UTR 3 NP_060154.3
NM_018080.3 12795 Intron NP_060550.2
NM_020821.2 12795 UTR 3 NP_065872.1
XM_011521713.2 12795 Intron XP_011520015.1
XM_011521714.2 12795 Intron XP_011520016.1

View Full Product Details