Product Details

SNP ID
rs1045466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1212203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGTCCGAGGGTTTCCGGGCCC[G/T]CTGCCCAGGCGCCGAGGCTGGAGAG
Phenotype
MIM: 602618 MIM: 605918
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CTBP1 PubMed Links

Gene Details

Gene
CTBP1
Gene Name
C-terminal binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012614.1 7471 UTR 3 NP_001012632.1
NM_001328.2 7471 UTR 3 NP_001319.1
XM_005272261.3 7471 UTR 3 XP_005272318.1
XM_005272263.4 7471 UTR 3 XP_005272320.1
XM_011513395.2 7471 UTR 3 XP_011511697.1
XM_011513396.2 7471 UTR 3 XP_011511698.1
XM_011513397.2 7471 UTR 3 XP_011511699.1
XM_017007762.1 7471 UTR 3 XP_016863251.1
XM_017007763.1 7471 UTR 3 XP_016863252.1
XM_017007764.1 7471 UTR 3 XP_016863253.1
XM_017007765.1 7471 UTR 3 XP_016863254.1
XM_017007766.1 7471 UTR 3 XP_016863255.1
XM_017007767.1 7471 UTR 3 XP_016863256.1
Gene
CTBP1-AS
Gene Name
CTBP1 antisense RNA
There are no transcripts associated with this gene.

Gene
LOC100130872
Gene Name
uncharacterized LOC100130872
There are no transcripts associated with this gene.

Gene
SPON2
Gene Name
spondin 2
There are no transcripts associated with this gene.

View Full Product Details