Product Details

SNP ID
rs1561889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65248488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGATACTCACATACATGGGCTTA[C/T]ACAGATGTGAATACTTACATACAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP16 PubMed Links

Gene Details

Gene
PARP16
Gene Name
poly(ADP-ribose) polymerase family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316943.1 Intron NP_001303872.1
NM_001316944.1 Intron NP_001303873.1
NM_017851.5 Intron NP_060321.3
XM_006720589.2 Intron XP_006720652.1
XM_006720590.2 Intron XP_006720653.1
XM_006720592.2 Intron XP_006720655.1
XM_011521742.2 Intron XP_011520044.1
XM_017022386.1 Intron XP_016877875.1
XM_017022387.1 Intron XP_016877876.1
XM_017022388.1 Intron XP_016877877.1
XM_017022389.1 Intron XP_016877878.1

View Full Product Details