Product Details

SNP ID
rs1063480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196051469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCTTTCAGGAATGAGGAAACC[A/T]GCTACATTCCTTATGGAAAGGCTTA
Phenotype
MIM: 190010
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TFRC PubMed Links

Gene Details

Gene
TFRC
Gene Name
transferrin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128148.2 2500 UTR 3 NP_001121620.1
NM_001313965.1 2500 UTR 3 NP_001300894.1
NM_001313966.1 2500 UTR 3 NP_001300895.1
NM_003234.3 2500 UTR 3 NP_003225.2

View Full Product Details