Product Details
- SNP ID
-
rs2583084
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:69468950 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGCTGCCTGGACAGGACTTTGACA[A/G]TATGGGACAATTCCCCTGAAAAAAA
- Phenotype
-
MIM: 610012
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SULF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75255789,rs79759599] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SULF1
- Gene Name
- sulfatase 1
View Full Product Details