Product Details

SNP ID
rs2236189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63244297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGACGCACCCCTGCCTTCCACCC[C/G]ACCCCACTCAACTGGGTAGGGGGCA
Phenotype
MIM: 605737 MIM: 612873
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BIRC7 PubMed Links

Gene Details

Gene
BIRC7
Gene Name
baculoviral IAP repeat containing 7
There are no transcripts associated with this gene.

Gene
FLJ16779
Gene Name
uncharacterized LOC100192386
There are no transcripts associated with this gene.

Gene
LOC107985425
Gene Name
uncharacterized LOC107985425
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017028201.1 Intron XP_016883690.1
Gene
MIR3196
Gene Name
microRNA 3196
There are no transcripts associated with this gene.

Gene
NKAIN4
Gene Name
Na+/K+ transporting ATPase interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152864.3 Intron NP_690603.3
XM_005260192.1 Intron XP_005260249.1
XM_005260194.3 Intron XP_005260251.1
XM_011528527.1 Intron XP_011526829.1
XM_011528528.2 Intron XP_011526830.1
XM_011528529.2 Intron XP_011526831.1
XM_017027636.1 Intron XP_016883125.1
XM_017027637.1 Intron XP_016883126.1
XM_017027638.1 Intron XP_016883127.1
XM_017027639.1 Intron XP_016883128.1

View Full Product Details