Product Details
- SNP ID
-
rs34041596
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63293891 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGGGAGGGGGCGTGTATTGGGGG[A/G]GTCTCTGTTCGGCTGCAGGGGGTGG
- Phenotype
-
MIM: 608377
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARFGAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs555485412] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARFGAP1
- Gene Name
- ADP ribosylation factor GTPase activating protein 1
There are no transcripts associated with this gene.
- Gene
- COL20A1
- Gene Name
- collagen type XX alpha 1 chain
- Gene
- MIR4326
- Gene Name
- microRNA 4326
There are no transcripts associated with this gene.
View Full Product Details