Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319956.1 | Intron | NP_001306885.1 | |||
NM_001319957.1 | Intron | NP_001306886.1 | |||
NM_018150.3 | Intron | NP_060620.2 | |||
XM_005270992.1 | Intron | XP_005271049.1 | |||
XM_005270993.1 | Intron | XP_005271050.1 | |||
XM_005270996.1 | Intron | XP_005271053.1 | |||
XM_006710735.3 | Intron | XP_006710798.1 | |||
XM_011541698.1 | Intron | XP_011540000.1 | |||
XM_011541699.1 | Intron | XP_011540001.1 | |||
XM_011541700.1 | Intron | XP_011540002.1 | |||
XM_011541702.1 | Intron | XP_011540004.1 | |||
XM_017001623.1 | Intron | XP_016857112.1 | |||
XM_017001624.1 | Intron | XP_016857113.1 | |||
XM_017001625.1 | Intron | XP_016857114.1 | |||
XM_017001626.1 | Intron | XP_016857115.1 |