Product Details

SNP ID
rs9393727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26499783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACACAGCACCTAAGGTGGACGGA[C/G]GCTCCAATAACGAAGATGGCTATAG
Phenotype
MIM: 601610
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BTN1A1 PubMed Links

Gene Details

Gene
BTN1A1
Gene Name
butyrophilin subfamily 1 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001732.2 494 Intron NP_001723.2
XM_005249340.3 494 UTR 5 XP_005249397.2

View Full Product Details