Product Details

SNP ID
rs11941012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:114828810 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCACCATCTCTCTTAAGCCCTCCT[A/C]AATATTTCAACACAAATGGTCTTTT
Phenotype
MIM: 615039
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NDST4 PubMed Links

Gene Details

Gene
NDST4
Gene Name
N-deacetylase and N-sulfotransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022569.2 Intron NP_072091.1
XM_017008545.1 Intron XP_016864034.1
XM_017008546.1 Intron XP_016864035.1

View Full Product Details