Product Details

SNP ID
rs11835716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55551474 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTCCCAGATTTCTGACCAGCA[C/T]GACAACAGGAAATAAAGTTATCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C4 PubMed Links

Gene Details

Gene
OR6C4
Gene Name
olfactory receptor family 6 subfamily C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005494.1 248 Missense Mutation ACG,ATG T83M NP_001005494.1

View Full Product Details