Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001104554.1 | Intron | NP_001098024.1 | |||
NM_017705.3 | Intron | NP_060175.3 | |||
XM_005254495.4 | Intron | XP_005254552.1 | |||
XM_011521720.2 | Intron | XP_011520022.1 | |||
XM_011521722.2 | Intron | XP_011520024.1 | |||
XM_011521723.2 | Intron | XP_011520025.1 | |||
XM_011521725.2 | Intron | XP_011520027.1 | |||
XM_011521727.2 | Intron | XP_011520029.1 | |||
XM_017022360.1 | Intron | XP_016877849.1 | |||
XM_017022361.1 | Intron | XP_016877850.1 | |||
XM_017022362.1 | Intron | XP_016877851.1 | |||
XM_017022363.1 | Intron | XP_016877852.1 | |||
XM_017022364.1 | Intron | XP_016877853.1 | |||
XM_017022365.1 | Intron | XP_016877854.1 | |||
XM_017022366.1 | Intron | XP_016877855.1 | |||
XM_017022367.1 | Intron | XP_016877856.1 | |||
XM_017022368.1 | Intron | XP_016877857.1 |