Product Details

SNP ID
rs3885685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:122321032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCCCTGAGACCGCCTCCCCAG[A/C]TTTTCTCCTGAAGCATTGTCTTGTG
Phenotype
MIM: 603779
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNCAIP PubMed Links

Gene Details

Gene
SNCAIP
Gene Name
synuclein alpha interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242935.2 Intron NP_001229864.1
NM_001308100.1 Intron NP_001295029.1
NM_001308105.1 Intron NP_001295034.1
NM_001308106.1 Intron NP_001295035.1
NM_001308107.1 Intron NP_001295036.1
NM_001308108.1 Intron NP_001295037.1
NM_001308109.1 Intron NP_001295038.1
NM_005460.3 Intron NP_005451.2
XM_005272138.3 Intron XP_005272195.1
XM_005272139.1 Intron XP_005272196.1
XM_006714734.2 Intron XP_006714797.1
XM_011543737.2 Intron XP_011542039.1
XM_011543738.2 Intron XP_011542040.1
XM_011543739.1 Intron XP_011542041.1
XM_011543741.2 Intron XP_011542043.1
XM_011543743.2 Intron XP_011542045.1
XM_011543745.1 Intron XP_011542047.1
XM_011543749.2 Intron XP_011542051.1
XM_011543750.1 Intron XP_011542052.1
XM_017010078.1 Intron XP_016865567.1
XM_017010079.1 Intron XP_016865568.1
XM_017010080.1 Intron XP_016865569.1
XM_017010081.1 Intron XP_016865570.1
XM_017010082.1 Intron XP_016865571.1
XM_017010083.1 Intron XP_016865572.1
XM_017010084.1 Intron XP_016865573.1
XM_017010085.1 Intron XP_016865574.1
XM_017010086.1 Intron XP_016865575.1

View Full Product Details