Product Details

SNP ID
rs3519
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112160517 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGATCTGTCTGGACCTCAATGT[A/G]CTCTCGGAGAAGCAGCCACGTTAGC
Phenotype
MIM: 613197
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HECTD4 PubMed Links

Gene Details

Gene
HECTD4
Gene Name
HECT domain E3 ubiquitin protein ligase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109662.3 15121 UTR 3 NP_001103132.3
Gene
MIR6861
Gene Name
microRNA 6861
There are no transcripts associated with this gene.

Gene
TRAFD1
Gene Name
TRAF-type zinc finger domain containing 1
There are no transcripts associated with this gene.

View Full Product Details