Product Details
- SNP ID
-
rs10256272
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:57465683 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAATTTACTAAGACTTATGAGTTC[C/T]AACAGAATGCACCAGGAGCAAATAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF716
PubMed Links
Gene Details
- Gene
- ZNF716
- Gene Name
- zinc finger protein 716
View Full Product Details