Product Details

SNP ID
rs9850589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120702882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGGGAAATTTTTCTGACTGAT[A/G]GAACTGTATCTCAAGACAGTGGGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RABL3 PubMed Links

Gene Details

Gene
RABL3
Gene Name
RAB, member of RAS oncogene family like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173825.3 Intron NP_776186.2
XM_006713595.2 Intron XP_006713658.1
XM_017006227.1 Intron XP_016861716.1
XM_017006228.1 Intron XP_016861717.1

View Full Product Details