Product Details

SNP ID
rs12743386
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:203866216 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAAAGGCAACATCTTAGAGATT[C/T]TAAGGATTTTAGGAGTTGTATGGGA
Phenotype
MIM: 128260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNRPE PubMed Links

Gene Details

Gene
SNRPE
Gene Name
small nuclear ribonucleoprotein polypeptide E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304464.1 Intron NP_001291393.1
NM_003094.3 Intron NP_003085.1

View Full Product Details