Product Details

SNP ID
rs35995150
Assay Type
Validated
NCBI dbSNP Submissions
35
Location
Chr.1:112722308 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CACTGTGCCTGGCCTGGCTGTGGAC[C/G]CACCTGACCTTGGCTGCCTTGCAGC
Phenotype
MIM: 609957
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM19A3 PubMed Links

Gene Details

Gene
FAM19A3
Gene Name
family with sequence similarity 19 member A3, C-C motif chemokine like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004440.1 431 Silent Mutation ACC,ACG T25T NP_001004440.1
NM_182759.2 431 Silent Mutation ACC,ACG T25T NP_877436.1
XM_005270766.3 431 Silent Mutation ACC,ACG T25T XP_005270823.1
XM_006710582.3 431 Silent Mutation ACC,ACG T25T XP_006710645.1
Gene
LOC105378910
Gene Name
uncharacterized LOC105378910
There are no transcripts associated with this gene.

Gene
PPM1J
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
There are no transcripts associated with this gene.

View Full Product Details