Product Details
- SNP ID
-
rs34861396
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:31640951 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCACTGGAGAAAGCCCAAGTTTA[A/G]TCCTGGGTTGGGGCCTCCTGAAACT
- Phenotype
-
MIM: 142445
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NRG1
PubMed Links
Gene Details
- Gene
- NRG1
- Gene Name
- neuregulin 1
View Full Product Details