Product Details

SNP ID
rs11785723
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:143692063 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGTCTGACCTCCCTGTGATGGCCA[C/T]GTGGGGTCCCAGGCCTGGTACTGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BREA2 PubMed Links

Gene Details

Gene
BREA2
Gene Name
breast cancer estrogen-induced apoptosis 2
There are no transcripts associated with this gene.

Gene
ZNF707
Gene Name
zinc finger protein 707
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100598.1 564 Intron NP_001094068.1
NM_001100599.1 564 Intron NP_001094069.1
NM_001288805.1 564 Intron NP_001275734.1
NM_001288806.1 564 Intron NP_001275735.1
NM_001288807.1 564 Intron NP_001275736.1
NM_001288808.1 564 Intron NP_001275737.1
NM_001288809.1 564 Intron NP_001275738.1
NM_173831.3 564 Intron NP_776192.2
XM_005250883.3 564 Intron XP_005250940.1
XM_006716542.3 564 Intron XP_006716605.1
XM_006716543.3 564 Intron XP_006716606.1
XM_011516977.2 564 Intron XP_011515279.1
XM_011516979.2 564 UTR 5 XP_011515281.1
XM_017013335.1 564 UTR 5 XP_016868824.1

View Full Product Details