Product Details

SNP ID
rs11043979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:18247102 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTTTAATATGCCAGTGTTCATTA[C/T]TGATGTCTAAGAGGAAAGTCTCCTA
Phenotype
MIM: 609001
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIK3C2G PubMed Links

Gene Details

Gene
PIK3C2G
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288772.1 Intron NP_001275701.1
NM_001288774.1 Intron NP_001275703.1
NM_004570.5 Intron NP_004561.3
XM_011520696.2 Intron XP_011518998.1
XM_011520697.2 Intron XP_011518999.1
XM_011520700.2 Intron XP_011519002.1
XM_011520701.2 Intron XP_011519003.1
XM_017019470.1 Intron XP_016874959.1
XM_017019471.1 Intron XP_016874960.1
XM_017019472.1 Intron XP_016874961.1
XM_017019473.1 Intron XP_016874962.1
XM_017019474.1 Intron XP_016874963.1
XM_017019475.1 Intron XP_016874964.1
XM_017019476.1 Intron XP_016874965.1
XM_017019477.1 Intron XP_016874966.1
XM_017019478.1 Intron XP_016874967.1
XM_017019479.1 Intron XP_016874968.1

View Full Product Details