Product Details

SNP ID
rs823088
Assay Type
Functionally tested
NCBI dbSNP Submissions
76
Location
Chr.1:205825047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTAGTGAATTCTTGCATGAAATA[C/T]TATCTACCAGTAGTGCTCAGTGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PM20D1 PubMed Links

Gene Details

Gene
PM20D1
Gene Name
peptidase M20 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152491.4 Intron NP_689704.4
XM_017000396.1 Intron XP_016855885.1
XM_017000397.1 Intron XP_016855886.1
XM_017000398.1 Intron XP_016855887.1

View Full Product Details