Product Details

SNP ID
rs10885634
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:114940677 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCACTTTATATAGTAGAGGCAGTA[C/T]GTCTGCAAGTTAGAAGGAGGTATAT
Phenotype
MIM: 610726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRUB1 PubMed Links

Gene Details

Gene
TRUB1
Gene Name
TruB pseudouridine synthase family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139169.4 Intron NP_631908.1

View Full Product Details