Product Details

SNP ID
rs36112041
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:14449919 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAATCGCATGACTTATTCTTATG[A/G]AGGGAATGTGGCAATATCTAACAAA
Phenotype
MIM: 604212
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PARN PubMed Links

Gene Details

Gene
PARN
Gene Name
poly(A)-specific ribonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134477.2 Intron NP_001127949.1
NM_001242992.1 Intron NP_001229921.1
NM_002582.3 Intron NP_002573.1
XM_011522510.2 Intron XP_011520812.1
XM_011522511.2 Intron XP_011520813.1
XM_011522513.2 Intron XP_011520815.1
XM_011522514.1 Intron XP_011520816.1
XM_017023258.1 Intron XP_016878747.1
XM_017023259.1 Intron XP_016878748.1
XM_017023260.1 Intron XP_016878749.1

View Full Product Details