Product Details

SNP ID
rs9556404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94602128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGCTGGCCCATCCCGCCGGCTG[A/G]GTCCGCCGGCCGCTCCTCCCGGCCC
Phenotype
MIM: 607787 MIM: 616146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR180 PubMed Links

Gene Details

Gene
GPR180
Gene Name
G protein-coupled receptor 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_180989.5 Intron NP_851320.1
Gene
TGDS
Gene Name
TDP-glucose 4,6-dehydratase
There are no transcripts associated with this gene.

View Full Product Details