Product Details

SNP ID
rs11770052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73861235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGTCAGATCCAGCCTTTTGGGGA[A/T]CCTGTTGCAGGTTACGAGGCTCTCA
Phenotype
MIM: 612547
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
WBSCR28 PubMed Links

Gene Details

Gene
WBSCR28
Gene Name
Williams-Beuren syndrome chromosome region 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182504.3 77 Missense Mutation AAC,ATC N14I NP_872310.2
XM_011515785.2 77 Intron XP_011514087.1
XM_017011741.1 77 Intron XP_016867230.1

View Full Product Details