Product Details

SNP ID
rs2233419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35402754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGAGCCTGACTCAGTGCGTCGG[A/G]GGCAGGAAGCACCAACCTGAGCATT
Phenotype
MIM: 164008
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFKBIA PubMed Links

Gene Details

Gene
NFKBIA
Gene Name
NFKB inhibitor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020529.2 Intron NP_065390.1

View Full Product Details