Product Details
- SNP ID
-
rs11722554
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:5015156 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCTCTGGTCTCAGTTCCCTTAGC[A/G]CTGACGATCTGGCAGGACCGTAGTC
- Phenotype
-
MIM: 607930
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYTL1
PubMed Links
Gene Details
- Gene
- CYTL1
- Gene Name
- cytokine like 1
View Full Product Details