Product Details

SNP ID
rs2842899
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:132538470 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTCATGATTGCTATCCTTCACTTC[A/T]AACAACTGCACACACCTACAAACTT
Phenotype
MIM: 608282
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TAAR9 PubMed Links

Gene Details

Gene
TAAR9
Gene Name
trace amine associated receptor 9 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175057.3 183 Nonsense Mutation AAA,TAA K61* NP_778227.3

View Full Product Details