Product Details

SNP ID
rs7546940
Assay Type
Functionally Tested
NCBI dbSNP Submissions
49
Location
Chr.1:196901662 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGGAAACTTTTTTTGGAAATATC[A/G]AGTATAAGCCTTATAATAACATAGT
Phenotype
MIM: 605337
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CFHR4 PubMed Links

Gene Details

Gene
CFHR4
Gene Name
complement factor H related 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201550.2 Intron NP_001188479.1
NM_001201551.1 Intron NP_001188480.1
NM_006684.4 Intron NP_006675.2
XM_006711129.3 Intron XP_006711192.1
XM_017000110.1 Intron XP_016855599.1
XM_017000111.1 Intron XP_016855600.1
XM_017000112.1 Intron XP_016855601.1
XM_017000113.1 Intron XP_016855602.1
XM_017000114.1 Intron XP_016855603.1

View Full Product Details