Product Details
- SNP ID
-
rs3766191
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
48
- Location
-
Chr.1:1082207 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- GCCAGGCATGGGGGTCCAGCGGATC[T/C]GAGCGGGGCCACAGAGCTCTGGAGG
- Phenotype
-
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf159
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12096277] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf159
- Gene Name
- chromosome 1 open reading frame 159
- Gene
- RNF223
- Gene Name
- ring finger protein 223
There are no transcripts associated with this gene.
View Full Product Details