Product Details

SNP ID
rs2804938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49157501 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGAATCACATAGACCAACAAGGA[A/G]AGAAGAAGAACAAAAAGAAACAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf128 PubMed Links

Gene Details

Gene
C10orf128
Gene Name
chromosome 10 open reading frame 128
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010863.2 Intron NP_001010863.1
NM_001288740.1 Intron NP_001275669.1
NM_001288741.1 Intron NP_001275670.1
NM_001288742.1 Intron NP_001275671.1
NM_001288743.1 Intron NP_001275672.1
XM_011539378.2 Intron XP_011537680.1
XM_011539379.1 Intron XP_011537681.1
XM_011539380.1 Intron XP_011537682.1
XM_011539382.1 Intron XP_011537684.1
XM_011539383.2 Intron XP_011537685.1
XM_011539384.1 Intron XP_011537686.1
XM_011539385.1 Intron XP_011537687.1
XM_017015792.1 Intron XP_016871281.1
XM_017015793.1 Intron XP_016871282.1
XM_017015794.1 Intron XP_016871283.1
XM_017015795.1 Intron XP_016871284.1
XM_017015796.1 Intron XP_016871285.1
XM_017015797.1 Intron XP_016871286.1
Gene
FAM170B-AS1
Gene Name
FAM170B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details