Product Details

SNP ID
rs12740822
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:111472670 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAGGCCCTTAATACAATGTGTCA[C/T]TGACTTATCATTTACTTGTTTTTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf162 PubMed Links

Gene Details

Gene
C1orf162
Gene Name
chromosome 1 open reading frame 162
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300834.1 Intron NP_001287763.1
NM_001300835.1 Intron NP_001287764.1
NM_174896.3 Intron NP_777556.1
XM_011540712.2 Intron XP_011539014.1
XM_017000323.1 Intron XP_016855812.1
XM_017000324.1 Intron XP_016855813.1

View Full Product Details