Product Details
- SNP ID
-
rs11900
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:194403692 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGAATCAAAATTACTATGTAATGGT[A/G]ACCTACGAGAGAAGACTCAGTCTAT
- Phenotype
-
MIM: 610232
MIM: 173511
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP13A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72607888] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP13A3
- Gene Name
- ATPase 13A3
- Gene
- GP5
- Gene Name
- glycoprotein V platelet
There are no transcripts associated with this gene.
View Full Product Details