Product Details
- SNP ID
-
rs6666245
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
38
- Location
-
Chr.1:26236408 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGTTGGGTGCTTTTTTTTTTTTT[C/T]CCCTCTGTCTGCCTCCATTCACTCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP85
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76374166] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP85
- Gene Name
- centrosomal protein 85
- Gene
- LOC101928303
- Gene Name
- uncharacterized LOC101928303
There are no transcripts associated with this gene.
View Full Product Details