Product Details

SNP ID
rs3128323
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:2023641 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGAGGCCACTGACTCAGGTCCCG[A/G]CCCCTCAACTCTCCTGGAGGGCTCG
Phenotype
MIM: 137163
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRD PubMed Links

Gene Details

Gene
GABRD
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000815.4 1912 Intron NP_000806.2
XM_011541194.2 1912 UTR 5 XP_011539496.1
XM_017000936.1 1912 UTR 5 XP_016856425.1
Gene
LOC105378589
Gene Name
uncharacterized LOC105378589
There are no transcripts associated with this gene.

View Full Product Details