Product Details

SNP ID
rs1418555
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:218350308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTTTGTACTCCCCCCAACACCC[A/G]CACCCCTACTCCCACTGGAACGTTT
Phenotype
MIM: 190220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGFB2 PubMed Links

Gene Details

Gene
TGFB2
Gene Name
transforming growth factor beta 2
There are no transcripts associated with this gene.

Gene
TGFB2-AS1
Gene Name
TGFB2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details