Product Details
- SNP ID
-
rs12759507
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
43
- Location
-
Chr.1:205577487 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGTGGAGCCAGCAGTGATGCTCG[C/T]GAGCATGTGGATCTCTGCTAACCCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MFSD4A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75126310] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MFSD4A
- Gene Name
- major facilitator superfamily domain containing 4A
View Full Product Details