Product Details

SNP ID
rs13128301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109829836 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTGTGTGTTGTTTGTGTTTCTGG[C/T]GGATCCACTGAAGGAAGCATAGCTG
Phenotype
MIM: 606468 MIM: 605224
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAR1 PubMed Links

Gene Details

Gene
GAR1
Gene Name
GAR1 ribonucleoprotein
There are no transcripts associated with this gene.

Gene
RRH
Gene Name
retinal pigment epithelium-derived rhodopsin homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006583.2 Intron NP_006574.1

View Full Product Details