Product Details

SNP ID
rs11731672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1405109 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACTCAGCCGGATTTGTTCGCCCT[C/T]CATCTCGGCCGACCCGGAAGAGGCC
Phenotype
MIM: 610203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRIPAK PubMed Links

Gene Details

Gene
CRIPAK
Gene Name
cysteine rich PAK1 inhibitor
There are no transcripts associated with this gene.

Gene
NKX1-1
Gene Name
NK1 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290079.1 Intron NP_001277008.1

View Full Product Details