Product Details

SNP ID
rs1136944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84139591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGAGGCGAAGGTGACCGGGGACC[G/T]AGGTACTGCTCCGGCTGGCCGGCGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GHITM PubMed Links

Gene Details

Gene
GHITM
Gene Name
growth hormone inducible transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014394.2 163 UTR 5 NP_055209.2

View Full Product Details