Product Details

SNP ID
rs2243903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127457464 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGCGGCAGCTGAGCGCCTGCTCC[C/T]TTTGGGCTGCCTCTTGCATGTCAGA
Phenotype
MIM: 610933 MIM: 180475
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRSAM1 PubMed Links

Gene Details

Gene
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005373.3 Intron NP_001005373.1
NM_001005374.3 Intron NP_001005374.1
NM_001190723.2 Intron NP_001177652.1
NM_138361.5 Intron NP_612370.3
XM_006717316.3 Intron XP_006717379.1
XM_017015283.1 Intron XP_016870772.1
XM_017015284.1 Intron XP_016870773.1
Gene
RPL12
Gene Name
ribosomal protein L12
There are no transcripts associated with this gene.

Gene
SNORA65
Gene Name
small nucleolar RNA, H/ACA box 65
There are no transcripts associated with this gene.

View Full Product Details